Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1B1 Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene
SDP

Catalog No. 502541386
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1386 30 μL
50-167-3705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1386 Supplier OriGene Supplier No. TA502667S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATP6V1B1 Monoclonal antibody specifically detects ATP6V1B1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen ATP6V1B1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2D2
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SULT1C2
Gene Accession No. O00338
Gene Alias ATP6B1; ATP6V1B1; ATPase H+ transporting V1 subunit B1; ATPase, H transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1 (Renal tubular acidosis with deafness); ATPase, H+ transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, V1 subunit B, isoform 1; AW208839; D630003L15; D630030L16Rik; D630039P21Rik; endomembrane proton pump 58 kDa subunit; H(+)-transporting two-sector ATPase, 58kD subunit; H+-ATPase beta 1 subunit; lysosomal 56/58kDa; RTA1B; vacuolar H+-ATPase; vacuolar proton pump 3; vacuolar proton pump subunit B 1; vacuolar proton pump, subunit 3; VATB; V-ATPase B1; V-ATPase B1 subunit; V-ATPase subunit B 1; VMA2; VPP3; Vpp-3; V-type proton ATPase subunit B, kidney isoform
Host Species Mouse
Immunogen Full length human recombinant protein of human SULT1C2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6819
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.