Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1B2 Antibody (OTI1E11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270238F
Description
ATP6V1B2 Monoclonal specifically detects ATP6V1B2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
ATP6V1B2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ATP6V1B2 | |
Full length recombinant protein of human ATP6V1B2 (NP_001684) produced in HEK293T cells. | |
0.1 mL | |
526.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI1E11 | |
Immunohistochemistry | |
000 subunit, 56/58kD, isoform 2, ATP6B2ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta polypeptide, ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2, Endomembrane proton pump 58 kDa subunit, H+ transporting two-sector ATPase, HO57ATP6B1B2, vacuolar H+-ATPase 56, Vacuolar proton pump subunit B 2, VATB, V-ATPase B2 subunit, V-ATPase subunit B 2, Vma2, VPP3ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 2, V-type proton ATPase subunit B, brain isoform | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction