Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1B2 Monoclonal Antibody (OTI1E11), TrueMAB™, OriGene
SDP

Catalog No. 502539825
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9825 30 μL
50-167-5764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9825 Supplier OriGene Supplier No. TA505244S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATP6V1B2 Monoclonal antibody specifically detects ATP6V1B2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ATP6V1B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZSCAN18
Gene Accession No. Q8TBC5
Gene Alias AI194269; AI790362; ATP6B1B2; Atp6b2; Atp6v1b2; atp6v1bb; atp6v1bb protein; ATPase H+ transporting V1 subunit B2; ATPase, H transporting, lysosomal V1 subunit B2; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 2; ATPase, H+ transporting, lysosomal 56/58kD, V1 subunit B, isoform 2; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2; ATPase, H+ transporting, lysosomal V1 subunit B2; ATPase, H+ transporting, lysosomal, V1 subunit B, member b; ATPase, H+ transporting, lysosomal, V1 subunit B2; ATPase, H+ transporting, V1 subunit B, isoform 2; DOOD; endomembrane proton pump 58 kDa subunit; fj51e01; H+ transporting two-sector ATPase; H+-ATPase B subunit; H+-ATPase non-catalytic subunit B; HO57; lysosomal 56/58kDa; R74844; testicular secretory protein Li 65; vacuolar H+-ATPase; vacuolar H+-ATPase 56,000 subunit; vacuolar H+ATPase B2; vacuolar proton pump subunit B 2; vacuolar-type H+ transporting ATPase subunit B2; Vat2; VATB; vatB2; V-ATPase B2 subunit; V-ATPase subunit B 2; Vma2; VPP3; V-type proton ATPase subunit B, brain isoform; wu:fj51e01; zgc:109771; ZLS2
Host Species Mouse
Immunogen Full length human recombinant protein of human ZSCAN18 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65982
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.