Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6V1B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577872 Supplier Invitrogen™ Supplier No. PA577872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS, Mouse brain, Rat brain Predicted Reactivity: Zebrafish (96%), Xenopus laevis (97%), Chicken (99%), Chimpanzee (99%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ATP6V1B2 encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A, three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. The protein encoded by this gene is one of two V1 domain B subunit isoforms and is the only B isoform highly expressed in osteoclasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V1B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Atp6v1b2
Gene Accession No. P21281, P62814, P62815
Gene Alias AI194269; AI790362; ATP6B1B2; Atp6b2; Atp6v1b2; atp6v1bb; atp6v1bb protein; ATPase H+ transporting V1 subunit B2; ATPase, H transporting, lysosomal V1 subunit B2; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 2; ATPase, H+ transporting, lysosomal 56/58kD, V1 subunit B, isoform 2; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2; ATPase, H+ transporting, lysosomal V1 subunit B2; ATPase, H+ transporting, lysosomal, V1 subunit B, member b; ATPase, H+ transporting, lysosomal, V1 subunit B2; ATPase, H+ transporting, V1 subunit B, isoform 2; DOOD; endomembrane proton pump 58 kDa subunit; fj51e01; H+ transporting two-sector ATPase; H+-ATPase B subunit; H+-ATPase non-catalytic subunit B; HO57; lysosomal 56/58kDa; R74844; testicular secretory protein Li 65; vacuolar H+-ATPase; vacuolar H+-ATPase 56,000 subunit; vacuolar H+ATPase B2; vacuolar proton pump subunit B 2; vacuolar-type H+ transporting ATPase subunit B2; Vat2; VATB; vatB2; V-ATPase B2 subunit; V-ATPase subunit B 2; Vma2; VPP3; V-type proton ATPase subunit B, brain isoform; wu:fj51e01; zgc:109771; ZLS2
Gene Symbols Atp6v1b2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ATP6V1B2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117596, 11966, 526
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.