Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1C1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | ATP6V1C1 |
---|---|
Dilution | Western Blot 1:5000-1:20000, Immunoprecipitation 1-5uL/mg of lysate |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ATP6V1C1 Polyclonal antibody specifically detects ATP6V1C1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ATP6V1C1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Endocrinology, Signal Transduction | |
0.2 um filtered solution in PBS | |
528 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:5000-1:20000, Immunoprecipitation 1-5uL/mg of lysate | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ATP6CH+-transporting ATPase chain C, vacuolar, ATP6Dsubunit C of vacuolar proton-ATPase V1 domain, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 42kD, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C, isoform 1, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C1, FLJ20057, H(+)-transporting two-sector ATPase, subunit C, vacuolar ATP synthase subunit C, vacuolar proton pump C subunit, Vacuolar proton pump subunit C 1, vacuolar proton pump, 42-kD subunit, vacuolar proton-ATPase, subunit C, VI domain, VATCH+ -ATPase C subunit, V-ATPase C subunit, V-ATPase subunit C 1, Vma5, V-type proton ATPase subunit C 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V1C1. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title