Learn More
Invitrogen™ ATP6V1C2 Monoclonal Antibody (OTI3E9)

Description
ATP6V1C2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ATP6V1C2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3E9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Atp6v1c2 |
| Gene Accession No. | Q8NEY4 |
| Gene Alias | 1110038G14Rik; Atp6c2; Atp6v1c2; ATPase H+ transporting V1 subunit C2; ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2; ATPase, H+ transporting, lysosomal V1 subunit C2; ATPase, H+ transporting, V1 subunit C, isoform 2; vacuolar H+ ATPase C2; vacuolar proton pump subunit C 2; V-ATPase C2 subunit; V-ATPase subunit C 2; VMA5; V-type ATPase C2 subunit a isoform; V-type proton ATPase subunit C 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.