Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1D Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297189PE
Description
ATP6V1D Polyclonal antibody specifically detects ATP6V1D in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ATP6V1D | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ATP6M, ATPase, H+ transporting lysosomal, member M, ATPase, H+ transporting, lysosomal (vacuolar proton pump), ATPase, H+ transporting, lysosomal 34kDa, V1 subunit D, H(+)-transporting two-sector ATPase, subunit M, vacuolar ATP synthase subunit D, vacuolar H-ATPase subunit D, vacuolar proton pump D subunit, vacuolar proton pump delta polypeptide, Vacuolar proton pump subunit D, vacuolar proton-ATPase subunit D, VATDV-ATPase D subunit, V-ATPase 28 kDa accessory protein, V-ATPase subunit D, VMA8, V-type proton ATPase subunit D | |
Produced in rabbits immunized with E. coli-derived Human ATP6V1D fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51382 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction