Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP6V1F Monoclonal Antibody (OTI5F10)
GREENER_CHOICE

Catalog No. PIMA525408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525408 Supplier Invitrogen™ Supplier No. MA525408
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATP6V1F Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V1F
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5F10
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Atp6v1f
Gene Accession No. P50408, Q16864
Gene Alias 1110004G16Rik; adenosinetriphosphatase 14k chain; ATP6S14; ATP6V1F; ATPase H+ transporting V1 subunit F; ATPase, H transporting, lysosomal V1 subunit F; ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F; ATPase, H+ transporting, lysosomal V1 subunit F; ATPase, H+ transporting, V1 subunit F; ATPase, vacuolar, 14 kD; H(+)-transporting two-sector ATPase, 14kD subunit; vacuolar ATP synthase subunit F; vacuolar proton pump F subunit; vacuolar proton pump subunit F; VATF; V-ATPase 14 kDa subunit; V-ATPase F subunit; V-ATPase subunit F; Vma7; V-type proton ATPase subunit F; v-type proton ATPase subunit F-like
Gene Symbols Atp6v1f
Host Species Mouse
Immunogen Full length human recombinant protein of ATP6V1F produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116664, 475199, 9296
Target Species Human, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.