Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1F Monoclonal Antibody (OTI6C11), TrueMAB™, OriGene
SDP

Catalog No. 502541634
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1634 30 μL
50-167-3385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1634 Supplier OriGene Supplier No. TA502322S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATP6V1F Monoclonal antibody specifically detects ATP6V1F in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen ATP6V1F
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI6C11
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDE2A
Gene Accession No. O00408
Gene Alias 1110004G16Rik; adenosinetriphosphatase 14k chain; ATP6S14; ATP6V1F; ATPase H+ transporting V1 subunit F; ATPase, H transporting, lysosomal V1 subunit F; ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F; ATPase, H+ transporting, lysosomal V1 subunit F; ATPase, H+ transporting, V1 subunit F; ATPase, vacuolar, 14 kD; H(+)-transporting two-sector ATPase, 14kD subunit; vacuolar ATP synthase subunit F; vacuolar proton pump F subunit; vacuolar proton pump subunit F; VATF; V-ATPase 14 kDa subunit; V-ATPase F subunit; V-ATPase subunit F; Vma7; V-type proton ATPase subunit F; v-type proton ATPase subunit F-like
Host Species Mouse
Immunogen Full length human recombinant protein of human PDE2A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5138
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.