Learn More
ATP6V1F Mouse anti-Canine, Human, Rat, Clone: OTI6C11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502322

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.Specifications
ATP6V1F | |
Monoclonal | |
Unconjugated | |
ATP6V1F | |
ATP6S14, VATF, Vma7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
116664, 475199, 9296 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI6C11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P50408, Q16864 | |
ATP6V1F | |
Full length human recombit protein of human ATP6V1F produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.