Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1G2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7248600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169319 25 μg
NB169318 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB169319 Supplier Novus Biologicals Supplier No. NBP31712125UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V1G2 Polyclonal antibody specifically detects ATP6V1G2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ATP6V1G2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias ATP6G2, ATPase, H+ transporting, lysosomal (vacuolar proton pump), ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 2, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G2, Em:AC004181.3, subunit G2, vacuolar ATP synthase subunit G 2, vacuolar proton pump G subunit 2, Vacuolar proton pump subunit G 2, V-ATPase 13 kDa subunit 2, V-ATPase subunit G 2, Vma10, V-type proton ATPase subunit G 2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: NLSAEVEQATRRQVQGMQSSQQRNRERVLAQLLGMVC
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 534
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.