Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1H Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232203C
Description
ATP6V1H Polyclonal antibody specifically detects ATP6V1H in Human, Mouse samples. It is validated for Western BlotSpecifications
ATP6V1H | |
Polyclonal | |
Western Blot | |
ATPase, H+ transporting, lysosomal 50/57kD, V1 subunit H, ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H, CGI-11, MSTP042, NBP1, Nef-binding protein 1, Protein VMA13 homolog, SFD, SFDalpha, SFDbeta, vacuolar ATP synthase subunit H, vacuolar ATPase subunit H, vacuolar proton pump H subunit, Vacuolar proton pump subunit H, Vacuolar proton pump subunit SFD, V-ATPase 50/57 kDa subunits, V-ATPase H subunit, V-ATPase subunit H, VMA13, V-type proton ATPase subunit H | |
The immunogen this antibody was made to, maps to a region between residue 400 to 450 of human ATPase, H+ Transporting, Lysosomal 50/57kDa, V1 Subunit H using the numbering given in entry NP_057025.2 (GeneID 51606). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
51606 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction