Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP7A Monoclonal Antibody (L60/4), PerCP
GREENER_CHOICE

Catalog No. PIMA545520
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545520 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545520 Supplier Invitrogen™ Supplier No. MA545520
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45520 was sufficient for detection of Copper-transporting ATPase1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat IgG:HRP as the secondary antibody. Detects approximately 180kDa in rat brain membrane preparations. This antibody was formerly sold as clone S60-4.

ATP7A (also known as Copper-transporting ATPase 1) functions as a transmembrane copper-translocating P-type ATPase and plays a vital role in systemic copper absorption in the gut and copper reabsorption in the kidney. Polarized epithelial cells such as Madin-Darby canine kidney cells are a physiologically relevant model for systemic copper absorption and reabsorption in vivo. Although ATP7A is not detectable in most normal tissues, it is expressed in a considerable fraction of many common tumor types. Increased expression of ATP7A renders cells resistant to cisplatin and carboplatin. Mutations in the ATP7A gene result in Menkes disease, which is fatal in early childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP7A
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone L60/4
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene ATP7A
Gene Accession No. P70705, Q04656, Q64430
Gene Alias ATP 7A; ATP7A; ATPase copper transporting alpha; ATPase, Cu++ transporting, alpha polypeptide; copper pump 1; Copper-transporting ATPase 1; Cu++-transporting P-type ATPase; DSMAX; FLJ17790; MC 1; MC1; Menke; Menkes disease-associated protein; menkes disease-associated protein homolog; Menkes protein; Menkes syndrome; MK; MNK; OHS; OTTHUMP00000062077; SMAX3
Gene Symbols ATP7A
Host Species Mouse
Immunogen Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11977, 24941, 538
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.