Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103110 Supplier Invitrogen™ Supplier No. PA5103110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATP7A.

ATP7A (also known as Copper-transporting ATPase 1) functions as a transmembrane copper-translocating P-type ATPase and plays a vital role in systemic copper absorption in the gut and copper reabsorption in the kidney. Polarized epithelial cells such as Madin-Darby canine kidney cells are a physiologically relevant model for systemic copper absorption and reabsorption in vivo. Although ATP7A is not detectable in most normal tissues, it is expressed in a considerable fraction of many common tumor types. Increased expression of ATP7A renders cells resistant to cisplatin and carboplatin. Mutations in the ATP7A gene result in Menkes disease, which is fatal in early childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP7A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATP7A
Gene Accession No. P70705, Q04656, Q64430
Gene Alias ATP 7A; ATP7A; ATPase copper transporting alpha; ATPase, Cu++ transporting, alpha polypeptide; copper pump 1; Copper-transporting ATPase 1; Cu++-transporting P-type ATPase; DSMAX; FLJ17790; MC 1; MC1; Menke; Menkes disease-associated protein; menkes disease-associated protein homolog; Menkes protein; Menkes syndrome; MK; MNK; OHS; OTTHUMP00000062077; SMAX3
Gene Symbols ATP7A
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP7A(Accession Q04656), corresponding to amino acid residues S1463-L1500.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11977, 24941, 538
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.