Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP7b Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100361AF405
Description
ATP7b Polyclonal specifically detects ATP7b in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ATP7b | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
P35670 | |
ATP7B | |
A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b. [UniProt# P35670] | |
Affinity Purified | |
RUO | |
Primary | |
Human and rat. Does not appear to work in mouse. | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease) | |
Rabbit | |
165 kDa | |
0.1 mL | |
Cancer, Signal Transduction | |
540 | |
Human, Rat, Mouse (Negative) | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction