Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP7b Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100361AF750
Description
ATP7b Polyclonal specifically detects ATP7b in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ATP7b | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease) | |
Rabbit | |
Affinity Purified | |
Cancer, Signal Transduction | |
540.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ATP7B | |
A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b. [UniProtNo. P35670] | |
0.1 mL | |
Primary | |
Human, Rat, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction