Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP7b Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 15 publications
$192.47 - $517.59
Specifications
| Antigen | ATP7b |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 10 ug/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunoblotting reported in scientific literature (PMID 29250175) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100360SS
![]() |
Novus Biologicals
NB100360SS |
0.025 mL |
Each for $192.47
|
|
|||||
NB100360
![]() |
Novus Biologicals
NB100360 |
0.1 mL |
Each for $517.59
|
|
|||||
Description
ATP7b Polyclonal specifically detects ATP7b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
| ATP7b | |
| Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 10 ug/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunoblotting reported in scientific literature (PMID 29250175) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease) | |
| ATP7B | |
| IgG | |
| Affinity Purified | |
| 165 kDa |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P35670 | |
| 540 | |
| A synthetic peptide made to an internal sequence near the N-terminus of human ATP7b. [UniProt# P35670] | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title