Learn More
Invitrogen™ ATP7B Monoclonal Antibody (3A12)

Description
Peptide Sequence: QLKCYKKPDL ERYEAQAHGH MKPLTASQVS VHIGMDDRWR DSPRATPWDQ VSYVSQVSLS SLTSDKPSRH SAAADDDGDK WSLLLNGRDE EQYI.
Specifications
Specifications
| Antigen | ATP7B |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3A12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Atp7b |
| Gene Accession No. | P35670 |
| Gene Alias | Atp7a; ATP7B; ATPase copper transporting beta; ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide (same as Wilson disease); copper pump 2; Copper-transporting ATPase 2; Hts; PINA; PINA gene, promoter; pineal night-specific ATPase; PWD; RP11-327P2.1; RP11-327P2.3; toxic milk; tx; WC1; WD; Wilson disease-associated protein; wilson disease-associated protein homolog; Wilson protein; Wnd; WND/140 kDa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.