Learn More
Invitrogen™ ATP7B Monoclonal Antibody (L62/29), PerCP

Description
1 μg/mL of MA5-45524 was sufficient for detection of Copper-transporting ATPase2 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat IgG:HRP as the secondary antibody. Detects approximately 160kDa in rat brain membrane preparations. This antibody was formerly sold as clone S62-29.
Specifications
Specifications
| Antigen | ATP7B |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | L62/29 |
| Concentration | 1 mg/mL |
| Conjugate | PerCP |
| Formulation | 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 |
| Gene | Atp7b |
| Gene Accession No. | P35670, Q64446, Q64535 |
| Gene Alias | Atp7a; ATP7B; ATPase copper transporting beta; ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide (same as Wilson disease); copper pump 2; Copper-transporting ATPase 2; Hts; PINA; PINA gene, promoter; pineal night-specific ATPase; PWD; RP11-327P2.1; RP11-327P2.3; toxic milk; tx; WC1; WD; Wilson disease-associated protein; wilson disease-associated protein homolog; Wilson protein; Wnd; WND/140 kDa |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.