Learn More
Invitrogen™ ATP7B Polyclonal Antibody, DyLight™ 488

Description
This antibody does not react with mouse samples.
Specifications
Specifications
| Antigen | ATP7B |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.93 mg/mL |
| Conjugate | DyLight 488 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | Atp7b |
| Gene Accession No. | P35670, Q64446, Q64535 |
| Gene Alias | Atp7a; ATP7B; ATPase copper transporting beta; ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide (same as Wilson disease); copper pump 2; Copper-transporting ATPase 2; Hts; PINA; PINA gene, promoter; pineal night-specific ATPase; PWD; RP11-327P2.1; RP11-327P2.3; toxic milk; tx; WC1; WD; Wilson disease-associated protein; wilson disease-associated protein homolog; Wilson protein; Wnd; WND/140 kDa |
| Gene Symbols | Atp7b |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.