Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP8 Mouse anti-Human, Unconjugated, Clone: 1G3-1H11-1C3, MilliporeSigma™

Catalog No. MABS21522MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS21522MI 25 μg
MABS21521MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS21522MI Supplier MilliporeSigma Supplier No. MABS215225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATP8 Monoclonal antibody specifically detects ATP8 in Human samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP8
Applications Western Blot
Classification Monoclonal
Clone 1G3-1H11-1C3
Conjugate Unconjugated
Gene Alias ATP synthase protein 8;A6L;F-ATPase subunit 8
Gene Symbols MT-ATP8:ATP8, ATPASE8, MTATP8
Host Species Mouse
Immunogen A synthetic peptide corresponding to the N-terminus of human mitochondrial ATP8.
Molecular Weight of Antigen ∼9.8 kDa observed; 8.0 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein L
Quantity 25 μg
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 1G3-1H11-1C3 is a mouse monoclonal antibody that specifically detects human mitochondrial ATPsynthase protein 8 (ATP8).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.