Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP8A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562707
Description
Immunogen sequence: KSQDPGAVVL GKSLTERAQL LKNVFKKNHV NLYRSESLQQ NLLHGYAFSQ DENGIVSQSE VI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The P-type adenosinetriphosphatases (P-type ATPases) are a family of proteins which use the free energy of ATP hydrolysis to drive uphill transport of ions across membranes. Several subfamilies of P-type ATPases have been identified. One subfamily catalyzes transport of heavy metal ions. Another subfamily transports non-heavy metal ions (NMHI). The protein encoded by this gene is a member of the third subfamily of P-type ATPases and acts to transport amphipaths, such as phosphatidylserine. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
ATP8A1 | |
Polyclonal | |
Unconjugated | |
ATP8A1 | |
AI481521; AI853962; aminophospholipid translocase; APLT; Atp3a2; Atp8a1; ATPase 8A1, aminophospholipid transporter (APLT), class I; ATPase 8A1, p type; ATPase class I type 8A member 1; ATPase II; ATPase phospholipid transporting 8A1; ATPase, aminophospholipid transporter (APLT), class I, type 8A, member 1; ATPASEII; Atpc1; ATPIA; ATPP2; AW743152; AW822227; B230107D19Rik; Chromaffin granule ATPase II; ClassI; P4-ATPase flippase complex alpha subunit ATP8A1; phospholipid-transporting ATPase IA; probable phospholipid-transporting ATPase IA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10396 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2Q0 | |
ATP8A1 | |
Recombinant protein corresponding to Human ATP8A1. Recombinant protein control fragment (Product #RP-100935). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction