Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP8A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558990 Supplier Invitrogen™ Supplier No. PA558990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLNGAGLDKA LKMSLPRRSR IRSSVGPVRS SLGYKKAEDE MSRATSVGDQ LEAPARTIYL NQPHLNKFRD NQISTAKYSV LTF Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 66%.

Catalytic component of a P4-ATPase flippase complex which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules. Reconstituted to liposomes, the ATP8A2:TMEM30A flippase complex predomiminantly transports phosphatidylserine (PS) and to a lesser extent phosphatidylethanolamine (PE). Proposed to function in the generation and maintenance of phospholipid asymmetry in photoreceptor disk membranes and neuronal axon membranes. May be involved in vesicle trafficking in neuronal cells. Involved in regulation of neurite outgrowth; acting in synergy with TMEM30A. Required for normal visual and auditory function; involved in photoreceptor and inner ear spiral ganglion cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP8A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP8A2
Gene Accession No. Q9NTI2
Gene Alias ATP; ATP8A2; ATPase class I type 8A member 2; ATPase phospholipid transporting 8A2; ATPase, aminophospholipid transporter, class I, type 8A, member 2; ATPase, aminophospholipid transporter-like, class I, type 8A, member 2; ATPIB; CAMRQ4; IB; ML-1; P4-ATPase flippase complex alpha subunit ATP8A2; Phospholipid-transporting ATPase IB; probable phospholipid-transporting ATPase IB
Gene Symbols ATP8A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP8A2. Recombinant protein control fragment (Product #RP-97955).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51761
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.