Learn More
Invitrogen™ ATP8A2 Polyclonal Antibody

Description
ATP8A2 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | ATP8A2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ATP8A2 |
| Gene Accession No. | Q9NTI2 |
| Gene Alias | ATP; ATP8A2; ATPase class I type 8A member 2; ATPase phospholipid transporting 8A2; ATPase, aminophospholipid transporter, class I, type 8A, member 2; ATPase, aminophospholipid transporter-like, class I, type 8A, member 2; ATPIB; CAMRQ4; IB; ML-1; P4-ATPase flippase complex alpha subunit ATP8A2; Phospholipid-transporting ATPase IB; probable phospholipid-transporting ATPase IB |
| Gene Symbols | ATP8A2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.