Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP8B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553839 Supplier Invitrogen™ Supplier No. PA553839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTFDEDSQPN DEVVPYSDDE TEDELDDQGS AVEPEQNRVN REAEENREPF RKECTWQVKA NDRKYHEQPH FMNTKFLCIK ESKYANNAIK TYKYNAFTFI PMNLFEQF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP8B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP8B1
Gene Accession No. O43520
Gene Alias ATP8B1; ATPase class I type 8B member 1; ATPase phospholipid transporting 8B1; ATPase, aminophospholipid transporter, class I, type 8B, member 1; ATPase, class I, type 8B, member 1; ATPIC; BRIC; E1-E2 ATPase; familial intrahepatic cholestasis type 1; FIC1; ICP1; P4-ATPase flippase complex alpha subunit ATP8B1; PFIC; PFIC1; phospholipid-transporting ATPase IC; probable phospholipid-transporting ATPase IC
Gene Symbols ATP8B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP8B1. Recombinant protein control fragment (Product #RP-92398).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.