Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP8B3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549285
Description
ATP8B3 Polyclonal Antibody for Western Blot
P4-ATPase flippase which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules. May be responsible for the maintenance of asymmetric distribution of phosphatidylserine (PS) in spermatozoa membranes. Involved in acrosome reactions and binding of spermatozoa to zona pellucida.
Specifications
ATP8B3 | |
Polyclonal | |
Unconjugated | |
ATP8B3 | |
aminophospholipid translocase ATP8B3; ATP1K; ATP8B3; ATPase class I type 8B member 3; ATPase phospholipid transporting 8B3; ATPase, aminophospholipid transporter, class I, type 8B, member 3; ATPase, class I, type 8B, member 3; ATPIK; FOS37502_2; Phospholipid-transporting ATPase IK; potential phospholipid-transporting ATPase IK; probable phospholipid-transporting ATPase IK | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
148229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O60423 | |
ATP8B3 | |
KLH conjugated synthetic peptide between 56-84 amino acids from the N-terminal region of human ATP8B3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction