Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP8B4 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305855UV
Description
ATP8B4 Polyclonal specifically detects ATP8B4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ATP8B4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
ATPase class I type 8B member 4, ATPase, class I, type 8B, member 4, ATPIM, EC 3.6.3, EC 3.6.3.1, FLJ25418, KIAA1939, potential phospholipid-transporting ATPase IM, probable phospholipid-transporting ATPase IM | |
Produced in rabbits immunized with E. coli-derived Human ATP8B4 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
79895 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction