Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP8B4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559122 Supplier Invitrogen™ Supplier No. PA559122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSINGRIYGE VHDDLDQKTE ITQEKEPVDF SVKSQADREF QFFDHNLMES IKMGDPKVHE FLR Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 73%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP8B4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP8B4
Gene Accession No. Q8TF62
Gene Alias ATP8B4; ATPase class I type 8B member 4; ATPase phospholipid transporting 8B4 (putative); ATPase, class I, type 8B, member 4; ATPIM; KIAA1939; P4-ATPase flippase complex alpha subunit ATP8B4; potential phospholipid-transporting ATPase IM; probable phospholipid-transporting ATPase IM
Gene Symbols ATP8B4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP8B4. Recombinant protein control fragment (Product #RP-98053).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.