Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP9A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597489 Supplier Invitrogen™ Supplier No. PA597489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP9A Polyclonal Antibody for ELISA, ICC/IF, IHC (P)

ATP9A, also named as ATPIIA and KIAA0611, belongs to the cation transport ATPase (P-type) family and Type IV subfamily. ATP9A catalyzes the reaction: ATP + H2O + phospholipid(In) = ADP + phosphate + phospholipid(Out).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP9A
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Atp9a
Gene Accession No. O75110
Gene Alias ATP9A; ATPase 9A, class II; ATPase 9A, p type; ATPase class II type 9A; ATPase phospholipid transporting 9A (putative); ATPase type IV, phospholipid-transporting (P-type),(putative); ATPase, class 2; ATPase, class II, type 9A; ATPIIA; IIa; KIAA0611; phospholipid-transporting ATPase IIA; probable phospholipid-transporting ATPase IIA
Gene Symbols Atp9a
Host Species Rabbit
Immunogen Recombinant Human Probable phospholipid-transporting ATPase IIA protein (2-69AA).
Purification Method Antigen affinity chromatography, Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10079
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.