Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ATPA
GREENER_CHOICESDP

Catalog No. 110750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-075-0 50 mg
11-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-075-0 Supplier R&D Systems™ Supplier No. 1107/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective GluK1 kainate agonist

A selective and potent GluK1 (formerly GluR5) kainate receptor agonist (Ki = 4.3 nM), inactive at GluK6 (formerly GluR6) (Ki > 1 mM) and only weakly active at AMPA receptors (GluA1-4) and the kainate receptors GluK5 (formerly KA-2) and GluK3 (formerly GluR7) (Ki values of 6 - 14 μM).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 140158-50-5
Molecular Formula C10H16N2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 228.25
Purity >98%
Molecular Weight (g/mol) 228.248
Chemical Name or Material ATPA
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.