Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATPAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565022 Supplier Invitrogen™ Supplier No. PA565022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFDLIWNRAQS CPTFLCALPR REGYEFFVGQ WTGTELHFTA LINIQTRGEA AASQLILYHY PELKEE Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.

This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 beta subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. Alternatively spliced transcript variants have been identified, but the biological validity of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATPAF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATPAF1
Gene Accession No. Q5TC12
Gene Alias 6330547J17Rik; AI593252; ATP synthase mitochondrial F1 complex assembly factor 1; ATP11; ATP11 homolog; ATP11p; ATPAF1; homolog of yeast ATP11
Gene Symbols ATPAF1
Host Species Rabbit
Immunogen Recombinant Human ATPAF1. Recombinant protein control fragment (Product #RP-105788).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.