Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATPAF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566257
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566257 Supplier Invitrogen™ Supplier No. PA566257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSASMSPGPTI PSPARAYAPP TERKRFYQNV SITQGEGGFE INLDHRKLKT PQAKLFTVPS EALAIAVATE WDSQ Highest antigen sequence identity to the following orthologs - mouse 87%, rat 87%.

This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 alpha subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. This gene is located within the Smith-Magenis syndrome region on chromosome 17. An alternatively spliced transcript variant has been described, but its biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATPAF2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Atpaf2
Gene Accession No. Q8N5M1
Gene Alias ATP synthase mitochondrial F1 complex assembly factor 2; ATP12; ATP12 homolog; ATP12p; ATPAF2; im:6893748; LP3663; MC5DN1
Gene Symbols Atpaf2
Host Species Rabbit
Immunogen Recombinant Human ATPAF2. Recombinant protein control fragment (Product #RP-106938).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91647
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.