Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Atpalpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532093 Supplier Invitrogen™ Supplier No. PA532093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Drosophila lysate. Predicted reactivity: Drosophila (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The ATPa, or ATPalpha gene, encodes a subunit of the Na+/K+ ATPase enzyme, which is crucial for maintaining cellular ion gradients by actively transporting Na+ out of cells and K+ into cells. This gene is essential in various organisms, including Drosophila melanogaster (fruit fly). Mutations in the ATPalpha gene can lead to significant developmental and physiological abnormalities, such as neurodegeneration, altered longevity, and myodegeneration. One of its critical roles is in neuromuscular functioning, where it affects locomotor activity, with mutations causing conditional bang-sensitive or temperature-sensitive paralysis. Additionally, ATPalpha mutations disrupt the formation and function of septate junctions in Drosophila, structures analogous to vertebrate tight junctions, further underscoring its role in maintaining cellular integrity and signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atpalpha
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Atpalpha
Gene Accession No. P13607
Gene Alias (Na[+] + K[+])-ATPase alpha-subunit; (Na[+],K[+])ATPase alpha subunit; (Na[+]/k[+])-ATPase; alpha-ATPase; alpha-subunit of ATPase; Atp; Atp a; ATPa; atpalpha; ATP-alpha; Atpalpha-PA; Atpalpha-PB; Atpalpha-PC; Atpalpha-PD; Atpalpha-PE; Atpalpha-PF; Atpalpha-PG; Atpalpha-PH; Atpalpha-PI; Atpalpha-PJ; Atpalpha-PK; ATPase alpha; ATPase alpha subunit; ATPBagr; CG5670; CG5670-PA; CG5670-PB; CG5670-PC; CG5670-PD; CG5670-PE; CG5670-PF; CG5670-PG; CG5670-PH; CG5670-PI; CG5670-PJ; CG5670-PK; Da-47; Dmel\CG5670; Dmel_CG5670; dnATPase; JY; K ATPase; K[+] -ATPase; K[+]-ATPase; K[+]-ATPase alpha subunit; K[+]-ATPase alpha-subunit; K-ATPase; l(3)01164; l(3)01453; l(3)04492; l(3)j3B2; l(3)j3B7; l(3)j6A4; l(3)rM454; l(3)ry136; Na; Na pump alpha subunit; Na(+)/K(+) ATPase alpha subunit; Na, K ATPase a subunit; Na,K ATPase; Na,K ATPase alpha subunit; Na,K-ATPase; Na,K-ATPase alpha; Na,K-ATPase alpha subunit; Na,K-ATPase alpha-subunit; Na/K ATPase; Na/K-ATPase; Na/K-ATPase alpha-subunit; Na[ +], K[ +] -ATPase; Na[+]; Na[+] pump alpha subunit; Na[+], K[+] ATPase alpha; Na[+],K[+] ATPase alpha subunit; Na[+],K[+]-ATPase alpha subunit; Na[+]/K[+] ATPase; Na[+]/K[+] ATPase (alpha subunit); Na[+]/K[+] ATPase alpha subunit; Na[+]/K[+]ATPase; Na[+]/K[+]-ATPase; Na[+]K[+] ATPase; Na[+]-K[+] ATPase; Na[+]K[+] ATPase alpha subunit; Na[+]K[+]ATPase; Na[+]K[+]-ATPase; Na[+]-K[+]-ATPase alpha[[5]]-subunit; Na+,K+ ATPase alpha-subunit; Na+/K+ ATPase alpha subunit; Na+/K+-ATPase; Na+-K+-ATPase; NaK; NaK ATPase; NaK-ATPase; Na-p; NKA; sodium pump alpha subunit; sodium pump alpha-subunit; Sodium pump subunit alpha; sodium/potassium transporting ATPase alpha; Sodium/potassium-transporting ATPase subunit alpha
Gene Symbols Atpalpha
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Drosophila Atpalpha. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 48971
Target Species Drosophila, Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.