Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATPase Inhibitory Factor 1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299177H
Description
ATPase Inhibitory Factor 1 Polyclonal specifically detects ATPase Inhibitory Factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATPase Inhibitory Factor 1 | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
ATP synthase inhibitor protein, ATPase inhibitor protein, ATPase inhibitor, mitochondrial, ATPase inhibitory factor 1, ATPIIF(1), ATPIP, IF1, Inhibitor of F(1)F(o)-ATPase, IP, MGC1167, MGC8898 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ATPase Inhibitory Factor 1. | |
0.1 mL | |
Lipid and Metabolism | |
93974 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction