Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATPase Na+/K+ beta 3 Rabbit anti-Human, Mouse, Rat, Clone: 0B7M7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | ATPase Na+/K+ beta 3 |
---|---|
Clone | 0B7M7 |
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
ATPase Na+/K+ beta 3 Monoclonal antibody specifically detects ATPase Na+/K+ beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ATPase Na+/K+ beta 3 | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ATPase, Na+/K+ transporting, beta 3 polypeptide, ATPB-3, CD298, CD298 antigen, FLJ29027, Na, K-ATPase beta-3 polypeptide, sodium/potassium-dependent ATPase beta-3 subunit, Sodium/potassium-dependent ATPase subunit beta-3, sodium/potassium-transporting ATPase beta-3 chain, sodium/potassium-transporting ATPase subunit beta-3 | |
A synthetic peptide corresponding to a sequence within amino acids 180-279 of human ATPase Na+/K+ beta 3 (P54709). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
0B7M7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, CD Markers, Endocrinology, Immunology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
483 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title