Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATPB Antibody (5F10), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326490100UL
Description
ATPB Monoclonal antibody specifically detects ATPB in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
ATPB | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
5F10 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit | |
A synthesized peptide derived from Human ATPB [UniProt P06576] | |
100 μL | |
Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
506 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction