Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATPBD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565952 Supplier Invitrogen™ Supplier No. PA565952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDEVEDLYELL KLVKEKEEVE GISVGAILSD YQRIRVENVC KRLNLQPLAY LWQRNQEDLL REMISSNIQA MIIKVAALGL DPDKHLG Highest antigen sequence identity to the following orthologs - mouse 93%, rat 65%.

Amidase that catalyzes the last step of diphthamide biosynthesis using ammonium and ATP. Diphthamide biosynthesis consists in the conversion of an L-histidine residue in the translation elongation factor (EEF2) to diphthamide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATPBD4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPH6
Gene Accession No. Q7L8W6
Gene Alias 5730421E18Rik; ATP binding domain 4; Atpbd4; ATP-binding domain-containing protein 4; diphthamide synthase; Diphthamide synthetase; diphthamine biosynthesis 6; diphthine--ammonia ligase; DPH6; DPH6 homolog; protein DPH6 homolog
Gene Symbols DPH6
Host Species Rabbit
Immunogen Recombinant Human ATPBD4. Recombinant protein control fragment (Product #RP-106641).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.