Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATPBD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571428 Supplier Invitrogen™ Supplier No. PA571428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATPBD4 Polyclonal Antibody for Western Blot

Amidase that catalyzes the last step of diphthamide biosynthesis using ammonium and ATP. Diphthamide biosynthesis consists in the conversion of an L-histidine residue in the translation elongation factor (EEF2) to diphthamide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATPBD4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DPH6
Gene Accession No. Q7L8W6
Gene Alias 5730421E18Rik; ATP binding domain 4; Atpbd4; ATP-binding domain-containing protein 4; diphthamide synthase; Diphthamide synthetase; diphthamine biosynthesis 6; diphthine--ammonia ligase; DPH6; DPH6 homolog; protein DPH6 homolog
Gene Symbols DPH6
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ATPBD4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89978
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.