Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATPG Antibody (OTI2H6), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270233B
Description
ATPG Monoclonal specifically detects ATPG in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ATPG | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ATP5F1C | |
Full length recombinant protein of human ATPG (NP_001001973) produced in E.coli. | |
0.1 mL | |
509.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2H6 | |
Western Blot, Immunohistochemistry | |
ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction