Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ATPgammaS tetralithium salt
GREENER_CHOICESDP

Catalog No. 408010 Shop All R&D Systems Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
40-801-0 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-801-0 Supplier Tocris Bioscience™ Supplier No. 4080/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-selective P2 agonist; analog of ATP (Cat. No. 3245)

ATPγS tetralithium salt is a P2 purinergic receptor agonist. Nonhydrolyzable analog of ATP (Cat. No. 3245).

Specifications

Quantity 10 mg
Product Type ATPγS tetralithium salt
Purity >90%
Molecular Weight (g/mol) 546.98

Product is intended for laboratory research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.