Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATPGD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558444 Supplier Invitrogen™ Supplier No. PA558444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMEFVEGTEH DVDLVLFGGR LLAAFVSDNG PTRLPGFTET AACMPTGLAP EQEAQMVQAA FRCCLGCGLL DGVFNVELKL TGAGPRLIEI NPRMGGFY Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

CARNS1 (EC 6.3.2.11), a member of the ATP-grasp family of ATPases, catalyzes the formation of carnosine (beta-alanyl-L-histidine) and homocarnosine (gamma-aminobutyryl-L-histidine), which are found mainly in skeletal muscle and the central nervous system, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATPGD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CARNS1
Gene Accession No. A5YM72
Gene Alias AI790298; ATPGD1; ATP-grasp domain containing 1; ATP-grasp domain-containing protein 1; Carnosine synthase 1; Carns1; Kiaa1394; mKIAA1394
Gene Symbols CARNS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATPGD1. Recombinant protein control fragment (Product #RP-97470).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57571
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.