Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atpif1 Mouse anti-Bovine, Human, Rat, Clone: 5E2D7, Abnova™

Catalog No. 89035404 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-404 Supplier Abnova Corporation Supplier No. MAB13999

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised agains recombinant rat Atpif1.

  • High specificity
  • High reactivity

Specifications

Antigen Atpif1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5E2D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised agains recombinant rat Atpif1.
Dilution Immunofluorescence Western Blot (1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Gene Accession No. Q03344
Gene Alias Atpi/IF1PA
Gene Symbols Atpif1
Host Species Mouse
Immunogen Recombinant protein corresponding to rat Atpif1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25392
Target Species Bovine, Human, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.