Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPIF1 Mouse anti-Human, Clone:1F3B8, Abnova™

Catalog No. 89334385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-385 Supplier Abnova Corporation Supplier No. MAB17937
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human ATPIF1.

This gene encodes a mitochondrial ATPase inhibitor. Alternative splicing occurs at this locus and three transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]

Specifications

Antigen ATPIF1
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F3B8
Conjugate Unconjugated
Description ATPase inhibitory factor 1
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunofluorescence (1:100-1:500)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ATPIF1
Gene Alias ATPI, ATPIP, IP, MGC1167, MGC8898
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human ATPIF1.
Quantity 100 μg
Gene ID (Entrez) 93974
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.