Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATPIF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592342
Description
Immunogen sequence: GSDQSENVDR GAGSIREAGG AFGKREQAEE ERYFRAQSRE QLAALKKHHE EEIVHHKKEI ERLQKEIERH KQKIKMLKHD D; Positive Samples: MCF-7, 22Rv1, HeLa, H460, Mouse brain; Cellular Location: Mitochondrion.
Complex V, also called F1F0 ATPase or ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP + phosphate to ATP, in oxidative phosphorylation, and as a proton pumping ATPase. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The enzyme was thought to be localized exclusively to mitochondria; however, it has recently been identified on the plasma membrane of several cell types including hepatocytes where it functions as the HDL receptor, on endothelial cells where it may act as the angiostatin receptor, and on the surface of cancer cells. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits; while the F0 consists of a, b, and c subunits. The enzyme in mammals is composed of 17 subunits, five of which make up the easily detached F1. The remainder subunits are components of two stalk domains and the proton pumping F0 part of the machinery. Two of the subunits of the F0 part are encoded on mitochondrial DNA while the other subunits are nuclearly encoded. Mutations in the mitochondrial-encoded subunits of ATP synthase (Complex V) cause OXPHOS disease.
Specifications
ATPIF1 | |
Polyclonal | |
Unconjugated | |
ATP5IF1 | |
ATP synthase F1 subunit epsilon; ATP synthase inhibitor protein; ATP synthase inhibitory factor subunit 1; Atp5if1; ATPase inhibitor (rat mitochondrial IF1 protein); ATPase inhibitor protein; ATPase inhibitor, mitochondrial; ATPase inhibitory factor 1; ATPase inhibitory factor 1, precursor; ATPI; Atpif1; ATPIP; IF(1); IF1; IF1PA; inhibitor of F(1)F(o)-ATPase; IP | |
Rabbit | |
Affinity chromatography | |
RUO | |
11983, 93974 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.93 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35143, Q9UII2 | |
ATP5IF1, Atpif1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 26-106 of human ATPIF1 (NP_0573951). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction