Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATR, p Thr1989 Antibody (222), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP243564

Catalog No. NBP243564


Only null left
Add to Cart

Description

Description

Ensure accurate, reproducible results in Immunoprecipitation

ATR Monoclonal specifically detects ATR in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

ATR (p Thr1989)
Monoclonal
Unconjugated
PBS, 20% Glycerol (pH7) with No Preservative
ATR
Carrier-protein conjugated synthetic peptide corresponding to residues around human ATR (phospho Thr1989). The exact sequence is proprietary.
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
222
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:100-1:500
ataxia telangiectasia and Rad3 related, Ataxia telangiectasia and Rad3-related protein, EC 2.7.11.1, FRAP-related protein 1, FRP1FRAP-related protein-1, MEC1, protein kinase ATR, Rad3 related protein, SCKL1, SCKLMEC1, mitosis entry checkpoint 1, homolog, serine/threonine-protein kinase ATR
Mouse
301 kDa
0.1 mL
Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, p53 Pathway, Virology Bacteria and Parasites
545
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only