Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATR Polyclonal Antibody, Invitrogen™

Catalog No. PIPA517265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517265 Supplier Thermo Scientific Supplier No. PA517265

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATR
Applications Western Blot
Classification Polyclonal
Concentration 14 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Gene ATR
Gene Accession No. Q13535
Gene Alias 2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8
Gene Symbols ATR
Host Species Rabbit
Immunogen Synthetic peptide corresponding to central residues of human ATR
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 545
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.