Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATRAID Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734957 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4957 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4957 Supplier Proteintech Group Inc Supplier No. 255481AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes all the three isoforms (20-24 kDa, 16-19 kDa and 28-30 kDa).

Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.

Specifications

Antigen ATRAID
Applications Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ATRAID
Gene Accession No. Q6PGD0, Q6UW56
Gene Alias Apoptosis related protein 3, APR 3, APR 3, APR3, C2orf28, HSPC013, p18, PRO240, UNQ214/PRO240
Gene Symbols ATRAID
Host Species Rabbit
Immunogen ATRAID Fusion Protein Ag22168
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381629, 51374
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.