Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atrial Natriuretic Peptide/ANP Antibody (2E3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Atrial Natriuretic Peptide/ANP |
---|---|
Clone | 2E3 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
Atrial Natriuretic Peptide/ANP Monoclonal antibody specifically detects Atrial Natriuretic Peptide/ANP in Human samples. It is validated for ELISASpecifications
Atrial Natriuretic Peptide/ANP | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4878 | |
IgG | |
Affinity purified |
2E3 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
ANF, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, PNDcardionatrin, prepronatriodilatin | |
A synthesized peptide derived from Human Atrial Natriuretic Peptide/ANP [UniProt P01160] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title