Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atrial Natriuretic Peptide/ANP Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP197752
Description
Atrial Natriuretic Peptide/ANP Polyclonal specifically detects Atrial Natriuretic Peptide/ANP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
proANP | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ANF, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, PNDcardionatrin, prepronatriodilatin | |
Goat | |
Affinity Purified | |
RUO | |
4878 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 0.03 ug/mL, Immunohistochemistry 5 - 10 ug/mL, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Peptide ELISA Detection limit 1:32000 | |
NP_006163.1 | |
NPPA | |
Peptide with sequence RIGAQSGLGCNSFR corresponding to C-Terminus according to NP_006163.1. | |
0.1 mg | |
Primary | |
The immunizing peptide represents aa137-150 of the precursor NPPA. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction