Learn More
Invitrogen™ Atrial Natriuretic Peptide (ANP) Competitive ELISA Kit
Description
The Atrial Natriuretic Peptide (ANP) Competitive ELISA quantitates ANP in human plasma,urine or cell culture medium.
Principle of the method
The Atrial Natriuretic Peptide (ANP) Competitive ELISA research-use-only kit is designed to quantitatively measure ANP present in plasma,urine,and tissue culture media samples. This kit is not recommended for serum or plasma samples without extraction. An ANP standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An ANP-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of an antibody to ANP to each well. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound ANP-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Assay Range | 0.741 to 180 ng/mL |
| Assay Sensitivity | 0.25 ng/mL |
| Description | Atrial Natriuretic Peptide (ANP) Competitive ELISA Kit |
| Assay | Multiplex assay on the Luminex Platform |
| Conjugate | HRP |
| Detection Method | Fluorescence |
| Product Type | ELISA |
| Protein Family | Peptide hormone |
| Sample Type | Plasma, Supernatant, Urine |
| Sufficient For | 96 assays |
| Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.